siRNA Details
| Gene Symbol: LINC01142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026957.1 | 3 | 397 | |||
| GenBank mRNA | BC033381.2 | - | 2 | 327 | ||
| DB058759.1 | - | 3 | 347 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG724047.1 | - | 3 | 351 | Chr. 1 - 170274031 - 170284157 [-] on Build GRCh38 | |

LINC01142
Genomic Map