siRNA Details
| Gene Symbol: SNX29P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045011.1 | 5 | 390 | |||
| GenBank mRNA | BC041583.1 | - | 2 | 254 | 4765274 | |
| DR003051.1 | - | 5 | 396 | |||
| Gene Symbol: SNX29P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002939.3 | 8 | 993 | |||
| GenBank mRNA | AK023827.1 | - | 9 | 1216 | ||
| BX648280.1 | - | 10 | 1665 | |||

SNX29P1
Genomic Map