siRNA Details
| Gene Symbol: CDIPT-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015396.1 | 3 | 280 | |||
| NR_024370.1 | 3 | 157 | ||||
| GenBank mRNA | AK131031.1 | - | 3 | 280 | ||
| BC070380.1 | - | 3 | 262 | |||
| BM006535.1 | - | 3 | 158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDIPT-AS1| Gene Symbol: CDIPT-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015396.1 | 3 | 280 | |||
| NR_024370.1 | 3 | 157 | ||||
| GenBank mRNA | AK131031.1 | - | 3 | 280 | ||
| BC070380.1 | - | 3 | 262 | |||
| BM006535.1 | - | 3 | 158 | |||