siRNA Details
| Gene Symbol: CDIPT-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015396.1 | 4 | 446 | |||
| NR_024370.1 | 4 | 323 | ||||
| GenBank mRNA | AK131031.1 | - | 4 | 446 | ||
| BC070380.1 | - | 4 | 428 | |||
| BM006535.1 | - | 4 | 324 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDIPT-AS1| Gene Symbol: CDIPT-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015396.1 | 4 | 446 | |||
| NR_024370.1 | 4 | 323 | ||||
| GenBank mRNA | AK131031.1 | - | 4 | 446 | ||
| BC070380.1 | - | 4 | 428 | |||
| BM006535.1 | - | 4 | 324 | |||