siRNA Details
Gene Symbol: YBX3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027011.1 | 1 | 1460 | |||
GenBank mRNA | DB520886.1 | - | 1 | 309 | ||
DB525052.1 | - | 1 | 174 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: YBX3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027011.1 | 1 | 1460 | |||
GenBank mRNA | DB520886.1 | - | 1 | 309 | ||
DB525052.1 | - | 1 | 174 |