siRNA Details
| Gene Symbol: YBX3P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027011.1 | 1 | 1428 | |||
| GenBank mRNA | DB520886.1 | - | 1 | 277 | ||
| DB525052.1 | - | 1 | 142 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

YBX3P1| Gene Symbol: YBX3P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027011.1 | 1 | 1428 | |||
| GenBank mRNA | DB520886.1 | - | 1 | 277 | ||
| DB525052.1 | - | 1 | 142 | |||