siRNA Details
| Gene Symbol: HERC2P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038215.2 | - | 2 | 303 | ||
| Gene Symbol: LOC102723753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135178.1 | 25 | 4376 | |||

HERC2P4
Genomic Map