siRNA Details
| Gene Symbol: GPR158-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027333.1 | 2 | 1691 | |||
| GenBank mRNA | AK094758.1 | - | 2 | 1692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR158-AS1| Gene Symbol: GPR158-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027333.1 | 2 | 1691 | |||
| GenBank mRNA | AK094758.1 | - | 2 | 1692 | ||