siRNA Details
| Gene Symbol: ESPNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026567.1 | 10 | 1710 | |||
| GenBank mRNA | AL035288.1 | - | 10 | 1710 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ESPNP| Gene Symbol: ESPNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026567.1 | 10 | 1710 | |||
| GenBank mRNA | AL035288.1 | - | 10 | 1710 | ||