siRNA Details
| Gene Symbol: LINC00176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027686.1 | 1 | 1646 | |||
| NR_027687.1 | 1 | 1646 | ||||
| GenBank mRNA | AL157500.1 | - | 1 | 327 | ||
| BC033342.1 | - | 1 | 1646 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00176| Gene Symbol: LINC00176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027686.1 | 1 | 1646 | |||
| NR_027687.1 | 1 | 1646 | ||||
| GenBank mRNA | AL157500.1 | - | 1 | 327 | ||
| BC033342.1 | - | 1 | 1646 | |||