siRNA Details
| Gene Symbol: LINC00176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027686.1 | 1 | 449 | |||
| NR_027687.1 | 1 | 449 | ||||
| GenBank mRNA | BC033342.1 | - | 1 | 449 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00176| Gene Symbol: LINC00176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027686.1 | 1 | 449 | |||
| NR_027687.1 | 1 | 449 | ||||
| GenBank mRNA | BC033342.1 | - | 1 | 449 | ||