siRNA Details
| Gene Symbol: LINC00472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026807.2 | 4 | 2789 | |||
| NR_121612.1 | 4 | 2840 | ||||
| NR_121613.1 | 3 | 2774 | ||||
| NR_121614.1 | 3 | 2723 | ||||
| GenBank mRNA | AK023251.1 | - | 4 | 2787 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00472| Gene Symbol: LINC00472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026807.2 | 4 | 2789 | |||
| NR_121612.1 | 4 | 2840 | ||||
| NR_121613.1 | 3 | 2774 | ||||
| NR_121614.1 | 3 | 2723 | ||||
| GenBank mRNA | AK023251.1 | - | 4 | 2787 | ||