siRNA Details
| Gene Symbol: LINC00942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028415.1 | 2 | 2018 | |||
| GenBank mRNA | BC019241.1 | - | 1 | 928 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00942| Gene Symbol: LINC00942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028415.1 | 2 | 2018 | |||
| GenBank mRNA | BC019241.1 | - | 1 | 928 | ||