siRNA Details
| Gene Symbol: LINC00494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026958.1 | 2 | 358 | |||
| GenBank mRNA | BC035164.2 | - | 2 | 321 | ||
| DB034461.1 | - | 2 | 358 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00494| Gene Symbol: LINC00494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026958.1 | 2 | 358 | |||
| GenBank mRNA | BC035164.2 | - | 2 | 321 | ||
| DB034461.1 | - | 2 | 358 | |||