siRNA Details
| Gene Symbol: LINC00494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026958.1 | 4 | 803 | |||
| GenBank mRNA | BC035164.2 | - | 4 | 766 | ||
| Gene Symbol: LINC01522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110027.1 | 3 | 595 | |||
| GenBank mRNA | DB088024.1 | - | 3 | 524 | ||

LINC00494
Genomic Map