siRNA Details
| Gene Symbol: RRN3P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003369.2 | 17 | 2160 | |||
| GenBank mRNA | AK225670.1 | - | 17 | 2010 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM686819.1 | - | 3 | 492 | Chr. 16 - 29104122 - 29116545 [+] on Build GRCh38 | |

RRN3P2
Genomic Map