siRNA Details
| Gene Symbol: GSTTP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003082.1 | 1 | 815 | |||
| GenBank mRNA | AK127991.1 | - | 1 | 815 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSTTP2| Gene Symbol: GSTTP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003082.1 | 1 | 815 | |||
| GenBank mRNA | AK127991.1 | - | 1 | 815 | ||