siRNA Details
| Gene Symbol: SUMO1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002189.3 | 1 | 724 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SUMO1P1| Gene Symbol: SUMO1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002189.3 | 1 | 724 | |||