siRNA Details
| Gene Symbol: HAR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_003245.1 | 2 | 850 | |||
| GenBank mRNA | DQ860410.1 | - | LIT3338 HAR1R |
2 | 850 | |
| DQ860411.1 | - | LIT3339 HAR1R |
3 | 727 | ||

HAR1B
Genomic Map