siRNA Details
| Gene Symbol: ZNF767P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027788.1 | 8 | 1386 | |||
| NR_027789.1 | 8 | 1362 | ||||
| GenBank mRNA | AK022762.1 | - | 8 | 1323 | ||
| AK124860.1 | - | 1 | 627 | |||
| AK295771.1 | - | 9 | 1348 | |||
| AY359125.1 | - | 5 | 547 | |||
| BC047675.1 | - | 8 | 1324 | 5727587 | ||

ZNF767P
Genomic Map