siRNA Details
| Gene Symbol: ZNF767P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027788.1 | 5 | 1050 | |||
| NR_027789.1 | 5 | 945 | ||||
| GenBank mRNA | AK022762.1 | - | 5 | 987 | ||
| AY359125.1 | - | 2 | 211 | |||
| BC047675.1 | - | 5 | 907 | 5727587 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF767P| Gene Symbol: ZNF767P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027788.1 | 5 | 1050 | |||
| NR_027789.1 | 5 | 945 | ||||
| GenBank mRNA | AK022762.1 | - | 5 | 987 | ||
| AY359125.1 | - | 2 | 211 | |||
| BC047675.1 | - | 5 | 907 | 5727587 | ||