siRNA Details
| Gene Symbol: ZNF767P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027788.1 | 6 | 1161 | |||
| NR_027789.1 | 6 | 1056 | ||||
| GenBank mRNA | AK022762.1 | - | 6 | 1098 | ||
| AK295771.1 | - | 7 | 1123 | |||
| AY359125.1 | - | 3 | 322 | |||
| BC047675.1 | - | 6 | 1018 | 5727587 | ||

ZNF767P
Genomic Map