siRNA Details
| Gene Symbol: ZNF767P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027788.1 | 8 | 3293 | |||
| NR_027789.1 | 9 | 1853 | ||||
| GenBank mRNA | AK022762.1 | - | 8 | 3230 | ||
| AK124860.1 | - | 1 | 2534 | |||
| AY359125.1 | - | 5 | 2454 | |||
| BC047675.1 | - | 9 | 1815 | 5727587 | ||

ZNF767P
Genomic Map