siRNA Details
| Gene Symbol: ZNF767P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027788.1 | 8 | 3565 | |||
| NR_027789.1 | 9 | 2125 | ||||
| GenBank mRNA | AK022762.1 | - | 8 | 3502 | ||
| AK124860.1 | - | 1 | 2806 | |||
| AY359125.1 | - | 5 | 2726 | |||
| BC047675.1 | - | 9 | 2087 | 5727587 | ||

ZNF767P
Genomic Map