siRNA Details
| Gene Symbol: TTTY14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF119903.1 | - | LIT2233 CYorf14 |
1 | 239 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI207481.1 | - | 1 | 239 | Chr. Y - 18877477 - 18878228 [-] on Build GRCh38 | |

TTTY14
Genomic Map