siRNA Details
| Gene Symbol: TTTY14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_125733.1 | 5 | 861 | |||
| NR_125734.1 | 4 | 781 | ||||
| NR_125735.1 | 3 | 706 | ||||
| NR_125736.1 | 5 | 456 | ||||
| GenBank mRNA | AF119903.1 | - | LIT2233 CYorf14 |
2 | 1223 | |
| BX280014.1 | - | 3 | 432 | |||
| HY121109.1 | - | 5 | 449 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY130111.1 | - | 5 | 449 | Chr. Y - 18872737 - 19075996 [-] on Build GRCh38 | |

TTTY14
Genomic Map