siRNA Details
| Gene Symbol: TTTY14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_125734.1 | 3 | 695 | |||
| NR_125735.1 | 2 | 620 | ||||
| NR_125736.1 | 4 | 370 | ||||
| GenBank mRNA | AF119903.1 | - | LIT2233 CYorf14 |
1 | 1137 | |
| BX280014.1 | - | 2 | 346 | |||
| FY211103.1 | - | 2 | 283 | |||
| FY211594.1 | - | 3 | 359 | |||
| HY121109.1 | - | 4 | 363 | |||

TTTY14
Genomic Map