siRNA Details
| Gene Symbol: LOC284798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027091.1 | 4 | 1245 | |||
| NR_027092.1 | 4 | 538 | ||||
| NR_027093.1 | 4 | 645 | ||||
| GenBank mRNA | AK095800.1 | - | 4 | 1245 | ||
| AK309371.1 | - | 4 | 538 | |||
| BC105718.1 | - | 4 | 645 | |||
| BC105722.1 | - | 4 | 645 | |||

LOC284798
Genomic Map