siRNA Details
| Gene Symbol: C20orf203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DB134998.1 | - | 1 | 240 | ||
| DB160076.1 | - | 1 | 122 | |||
| GU931819.1 | - | 1 | 240 | |||
| GU931820.1 | - | 1 | 240 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C20orf203| Gene Symbol: C20orf203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DB134998.1 | - | 1 | 240 | ||
| DB160076.1 | - | 1 | 122 | |||
| GU931819.1 | - | 1 | 240 | |||
| GU931820.1 | - | 1 | 240 | |||