siRNA Details
| Gene Symbol: PLEKHM1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024386.2 | 11 | 2577 | |||
| GenBank mRNA | AL833315.1 | - | 11 | 2547 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA421664.1 | - | 4 | 522 | Chr. 17 - 64785129 - 64792470 [-] on Build GRCh38 | |

PLEKHM1P1
Genomic Map