siRNA Details
| Gene Symbol: LOC440461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027283.1 | 1 | 1294 | |||
| GenBank mRNA | BC080624.1 | - | 1 | 1118 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC440461| Gene Symbol: LOC440461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027283.1 | 1 | 1294 | |||
| GenBank mRNA | BC080624.1 | - | 1 | 1118 | ||