siRNA Details
| Gene Symbol: BAIAP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026857.1 | 2 | 2217 | |||
| GenBank mRNA | AK027350.1 | - | 1 | 81 | ||
| AK056555.1 | - | 2 | 2199 | |||
| AK075238.1 | - | 1 | 180 | |||
| AK096609.1 | - | 1 | 1125 | |||
| AK124762.1 | - | 2 | 2258 | |||
| BC110822.1 | - | 1 | 226 | |||

BAIAP2-AS1
Genomic Map