siRNA Details
| Gene Symbol: BAIAP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026857.1 | 2 | 1880 | |||
| GenBank mRNA | AK056555.1 | - | 2 | 1862 | ||
| AK096609.1 | - | 1 | 788 | |||
| AK124762.1 | - | 2 | 1921 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BAIAP2-AS1| Gene Symbol: BAIAP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026857.1 | 2 | 1880 | |||
| GenBank mRNA | AK056555.1 | - | 2 | 1862 | ||
| AK096609.1 | - | 1 | 788 | |||
| AK124762.1 | - | 2 | 1921 | |||