siRNA Details
| Gene Symbol: ANP32A-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026808.1 | 1 | 2189 | |||
| GenBank mRNA | AK021784.1 | - | 1 | 2189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANP32A-IT1| Gene Symbol: ANP32A-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026808.1 | 1 | 2189 | |||
| GenBank mRNA | AK021784.1 | - | 1 | 2189 | ||