siRNA Details
Gene Symbol: AATBC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026961.1 | 1 | 2695 | |||
GenBank mRNA | AK128598.1 | - | 1 | 2696 | ||
BC111558.1 | - | 1 | 2694 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000400385.2 | - | 1 | 2695 | Chr. 21 - 43805758 - 43812567 [-] on Build GRCh38 |
