siRNA Details
| Gene Symbol: AATBC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026961.1 | 2 | 4336 | |||
| GenBank mRNA | AK093410.1 | - | 4 | 1627 | ||
| AK128598.1 | - | 2 | 4337 | |||
| BC034348.1 | - | 2 | 670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AATBC| Gene Symbol: AATBC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026961.1 | 2 | 4336 | |||
| GenBank mRNA | AK093410.1 | - | 4 | 1627 | ||
| AK128598.1 | - | 2 | 4337 | |||
| BC034348.1 | - | 2 | 670 | |||