siRNA Details
| Gene Symbol: CLEC4GP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002931.2 | 7 | 1267 | |||
| GenBank mRNA | AK090689.1 | - | 7 | 1249 | ||
| AK090734.1 | - | 7 | 1251 | |||
| AK123140.1 | - | 7 | 1233 | |||
| BC047593.1 | - | 5 | 1141 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLEC4GP1| Gene Symbol: CLEC4GP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002931.2 | 7 | 1267 | |||
| GenBank mRNA | AK090689.1 | - | 7 | 1249 | ||
| AK090734.1 | - | 7 | 1251 | |||
| AK123140.1 | - | 7 | 1233 | |||
| BC047593.1 | - | 5 | 1141 | |||