siRNA Details
| Gene Symbol: CLEC4GP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002931.2 | 6 | 654 | |||
| GenBank mRNA | AK090689.1 | - | 6 | 654 | ||
| AK090734.1 | - | 6 | 651 | |||
| AK123140.1 | - | 6 | 635 | |||
| BC047593.1 | - | 4 | 543 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG704308.1 | - | 4 | 548 | Chr. 19 - 7788377 - 7789908 [+] on Build GRCh38 | |

CLEC4GP1
Genomic Map