siRNA Details
Gene Symbol: LINC00574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026780.1 | 3 | 2265 | |||
GenBank mRNA | AK023224.1 | - | 3 | 2017 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000420557.2 | - | 3 | 2017 | Chr. 6 - 169790321 - 169802873 [+] on Build GRCh38 |
