siRNA Details
| Gene Symbol: LINC00574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026780.1 | 1 | 96 | |||
| GenBank mRNA | DA918406.1 | - | 1 | 96 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB259408.1 | - | 1 | 103 | Chr. 6 - 169790066 - 169800892 [+] on Build GRCh38 | |

LINC00574
Genomic Map