siRNA Details
| Gene Symbol: LINC00574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026780.1 | 3 | 1082 | |||
| GenBank mRNA | AK023224.1 | - | 3 | 834 | ||
| BC101251.2 | - | 2 | 622 | |||
| BC101252.1 | - | 2 | 622 | |||
| BC101253.1 | - | 2 | 622 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00574| Gene Symbol: LINC00574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026780.1 | 3 | 1082 | |||
| GenBank mRNA | AK023224.1 | - | 3 | 834 | ||
| BC101251.2 | - | 2 | 622 | |||
| BC101252.1 | - | 2 | 622 | |||
| BC101253.1 | - | 2 | 622 | |||