siRNA Details
| Gene Symbol: NOP14-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015453.2 | 4 | 3480 | |||
| GenBank mRNA | AB000464.1 | - | 4 | 3480 | ||
| AW134854.1 | - | 1 | 72 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOP14-AS1| Gene Symbol: NOP14-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015453.2 | 4 | 3480 | |||
| GenBank mRNA | AB000464.1 | - | 4 | 3480 | ||
| AW134854.1 | - | 1 | 72 | |||