siRNA Details
| Gene Symbol: TMEM254-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027428.1 | 6 | 3716 | |||
| NR_027429.1 | 5 | 3629 | ||||
| NR_027430.1 | 5 | 3505 | ||||
| NR_027431.2 | 5 | 3479 | ||||
| NR_027432.1 | 4 | 3392 | ||||
| GenBank mRNA | BX094741.1 | - | 1 | 152 | ||

TMEM254-AS1
Genomic Map