siRNA Details
| Gene Symbol: TMEM254-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027428.1 | 6 | 3403 | |||
| NR_027429.1 | 5 | 3316 | ||||
| NR_027430.1 | 5 | 3192 | ||||
| NR_027431.2 | 5 | 3166 | ||||
| NR_027432.1 | 4 | 3079 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM254-AS1| Gene Symbol: TMEM254-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027428.1 | 6 | 3403 | |||
| NR_027429.1 | 5 | 3316 | ||||
| NR_027430.1 | 5 | 3192 | ||||
| NR_027431.2 | 5 | 3166 | ||||
| NR_027432.1 | 4 | 3079 | ||||