siRNA Details
| Gene Symbol: FAM86C2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024249.1 | 4 | 1075 | |||
| GenBank mRNA | AK055771.1 | - | 2 | 1728 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM86C2P| Gene Symbol: FAM86C2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024249.1 | 4 | 1075 | |||
| GenBank mRNA | AK055771.1 | - | 2 | 1728 | ||