siRNA Details
| Gene Symbol: MIR765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030527.1 | 1 | 104 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR765| Gene Symbol: MIR765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030527.1 | 1 | 104 | |||