siRNA Details
| Gene Symbol: MIR765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030527.1 | 1 | 41 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA054058.1 | - | 2 | 387 | Chr. 1 - 156936135 - 156991697 [-] on Build GRCh38 | |

MIR765
Genomic Map