siRNA Details
| Gene Symbol: BIN3-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027715.1 | 1 | 752 | |||
| GenBank mRNA | AK024169.1 | - | 1 | 705 | ||
| BC013100.1 | - | 1 | 348 | 4389733 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BIN3-IT1| Gene Symbol: BIN3-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027715.1 | 1 | 752 | |||
| GenBank mRNA | AK024169.1 | - | 1 | 705 | ||
| BC013100.1 | - | 1 | 348 | 4389733 | ||