siRNA Details
Gene Symbol: BIN3-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027715.1 | 1 | 101 | |||
GenBank mRNA | AK024169.1 | - | 1 | 54 | ||
DN996902.1 | - | 1 | 109 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BIN3-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027715.1 | 1 | 101 | |||
GenBank mRNA | AK024169.1 | - | 1 | 54 | ||
DN996902.1 | - | 1 | 109 |