siRNA Details
| Gene Symbol: LINC00982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015440.1 | 1 | 60 | |||
| NR_024371.1 | 1 | 60 | ||||
| GenBank mRNA | BC029785.1 | - | 1 | 104 | ||
| BM931404.1 | - | 1 | 61 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00982| Gene Symbol: LINC00982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015440.1 | 1 | 60 | |||
| NR_024371.1 | 1 | 60 | ||||
| GenBank mRNA | BC029785.1 | - | 1 | 104 | ||
| BM931404.1 | - | 1 | 61 | |||